Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUC1 SIRNA Assays ›
Gene Symbol:
MUC1
Gene Name:
mucin 1, cell surface associated
Gene Aliases:
ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM
Chromosome Location:
Chr.1: 155185824 - 155192915 on Build GRCh38
UniGene ID:
Hs.89603
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
262783
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001018016.2
NP_001018016.1
3
292
NM_001018017.2
NP_001018017.1
3
265
NM_001044390.2
NP_001037855.1
3
246
NM_001044391.2
NP_001037856.1
3
265
NM_001044392.2
NP_001037857.1
3
292
NM_001044393.2
NP_001037858.1
3
246
NM_001204285.1
NP_001191214.1
3
925
NM_001204286.1
NP_001191215.1
3
952
NM_001204287.1
NP_001191216.1
3
346
NM_001204288.1
NP_001191217.1
3
292
NM_001204292.1
NP_001191221.1
3
273
NM_001204293.1
NP_001191222.1
3
319
NM_001204294.1
NP_001191223.1
3
246
NM_001204295.1
NP_001191224.1
3
292
NM_001204296.1
NP_001191225.1
3
273
NM_001204297.1
NP_001191226.1
3
346
NM_002456.5
NP_002447.4
3
319
GenBank mRNA
AF125525.1
-
3
184
AF348143.1
-
3
271
AK301575.1
-
4
1177
AY327582.1
-
3
198
AY327583.1
-
3
217
AY327584.1
-
3
244
AY327587.1
-
3
270
AY327590.1
-
3
198
AY327591.1
-
3
198
AY327592.1
-
3
217
AY327595.1
-
3
217
AY327596.1
-
4
400
AY327597.1
-
3
198
AY327598.1
-
3
198
AY327599.1
-
3
225
AY327600.1
-
3
217
AY466157.1
-
3
174
BC120974.2
-
3
242
40115473
BC120975.2
-
3
269
40115475
EF583642.1
-
3
595
EF583643.1
-
3
423
EF583644.1
-
3
409
EF583645.1
-
3
369
EF583647.1
-
3
324
EF583648.1
-
3
251
EF583649.1
-
3
270
EF583650.1
-
3
207
EF583651.1
-
3
207
EF583655.1
-
3
270
EF670706.1
-
3
619
EF670707.1
-
3
288
EF670709.1
-
3
251
EF670710.1
-
3
243
EF670711.1
-
3
297
EF670712.1
-
3
224
EF670713.1
-
3
270
EF670716.1
-
3
270
EF670717.1
-
3
270
EF670718.1
-
3
324
EF670719.1
-
3
294
EF670721.1
-
3
251
FJ226028.1
-
2
726
FJ226029.1
-
2
884
FJ226030.1
-
3
854
FJ226031.1
-
2
812
FJ226032.1
-
2
745
FJ226033.1
-
2
726
FJ226034.1
-
3
645
FJ226036.1
-
2
745
FJ226037.1
-
3
423
FJ226038.1
-
2
744
FJ226040.1
-
3
270
FJ226042.1
-
3
324
FJ226043.1
-
3
310
FJ226044.1
-
3
350
FJ226047.1
-
3
224
FJ226049.1
-
3
324
FJ226050.1
-
3
297
FJ226052.1
-
3
243
FJ226053.1
-
3
251
FJ226054.1
-
3
369
FJ226055.1
-
3
251
J05581.1
-
3
925
J05582.1
-
3266
M34088.1
-
1
412
M34089.1
-
1
412
U60259.1
-
3
199
U60260.1
-
3
218
U60261.1
-
3
272
X52228.1
-
4
1030
X52229.1
-
4
1030
X80761.1
-
3
193
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
negative regulation of transcription by competitive promoter binding
O-glycan processing
negative regulation of cell adhesion mediated by integrin
positive regulation of transcription from RNA polymerase II promoter in response to stress
regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of histone H4 acetylation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
RNA polymerase II core promoter proximal region sequence-specific DNA binding
p53 binding
transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0