siRNA Details
Gene Symbol: Snrpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082961.1 | NP_001076430.1 | 6, 7 | 734 | ||
NM_001082962.1 | NP_001076431.1 | 6, 7 | 778 | |||
NM_013670.3 | NP_038698.1 | 5, 6 | 672 | |||
GenBank mRNA | AK010671.1 | - | 5, 6 | 709 | ||
AK013607.1 | - | 5, 6 | 629 | |||
AK144386.1 | - | 5, 6 | 628 | |||
BC019589.1 | - | 5, 6 | 620 | 3484323 | ||
BC024880.1 | - | 5, 6 | 611 | 3987218 | ||
BC069894.1 | - | 5, 6 | 609 | |||
S40304.1 | - | 2, 3 | 317 | |||
S62288.1 | - | 2, 3 | 194 | |||
X62648.1 | - | 2, 3 | 317 | |||
X63730.1 | - | 2, 3 | 270 |
Gene Symbol: Snurf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033174.3 | NP_149409.1 | 5, 6 | 672 | ||
GenBank mRNA | CF751578.1 | - | 1, 2 | 171 |

More Information
Other Species Targeted

Panther Classification:
Molecular Function -




