Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146986.1 | NP_667197.1 | 1 | 740 | ||
GenBank mRNA | BC106817.1 | - | 1 | 739 | ||
BC128054.1 | - | 1 | 740 | 40043588 |