Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012401.3 | NP_036533.2 | 35 | 5477 | ||
XM_005261909.1 | XP_005261966.1 | 36 | 5525 | |||
XM_005261910.2 | XP_005261967.1 | 36 | 5536 | |||
XM_005261911.4 | XP_005261968.1 | 35 | 5475 | |||
XM_006724413.2 | XP_006724476.1 | 37 | 5595 | |||
XM_011530682.2 | XP_011528984.1 | 36 | 5676 | |||
XM_017028703.1 | XP_016884192.1 | 37 | 5576 | |||
XM_017028704.1 | XP_016884193.1 | 36 | 5529 | |||
GenBank mRNA | AB002313.1 | - | 34 | 5394 | ||
AB075511.1 | - | 20 | 2587 | |||
AK025415.1 | - | 10 | 1064 | |||
AK025701.1 | - | 22 | 2845 | |||
AK056543.1 | - | 18 | 2646 | |||
AK074932.1 | - | 17 | 2113 | |||
AK123131.1 | - | 7 | 1058 | |||
AK126394.1 | - | 16 | 2186 | |||
BC004542.2 | - | 12 | 1296 | |||
BC110366.1 | - | 7 | 714 | |||
BT006887.1 | - | 12 | 1280 |