Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018117.11 | NP_060587.8 | 3 | 466 | ||
XM_005269962.2 | XP_005270019.1 | 3 | 183 | |||
XM_005269963.2 | XP_005270020.1 | 3 | 315 | |||
XM_017016397.1 | XP_016871886.1 | 3 | 328 | |||
XM_017016398.1 | XP_016871887.1 | 3 | 258 | |||
XM_017016399.1 | XP_016871888.1 | 3 | 258 | |||
XM_017016400.1 | XP_016871889.1 | 3 | 357 | |||
GenBank mRNA | AB037772.2 | - | 3 | 279 | ||
AF320223.1 | - | 3 | 253 | |||
AK301761.1 | - | 3 | 249 | |||
AK315145.1 | - | 3 | 244 | |||
AL137699.1 | - | 2 | 42 | |||
BC040469.1 | - | 3 | 283 | 4730308 | ||
BC071564.1 | - | 3 | 249 | 30346203 | ||
BI912281.1 | - | 3 | 486 | |||
DC342128.1 | - | 3 | 466 |