Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080442.2 | NP_001073911.1 | 9 | 1202 | ||
XM_017022946.1 | XP_016878435.1 | 11 | 1667 |