Search
Search

KCTD19| Gene Symbol: KCTD19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100915.1 | NP_001094385.1 | 3, 4 | 501 | ||
| GenBank mRNA | AK097481.1 | - | 3, 4 | 504 | ||
| AK302567.1 | - | 3, 4 | 496 | |||
| AK302748.1 | - | 3, 4 | 485 | |||
| BC070103.1 | - | 3, 4 | 469 | |||
| BC094790.1 | - | 3, 4 | 472 | |||
| BC117335.1 | - | 3, 4 | 501 | 40125886 | ||