Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319099.1 | NP_001306028.1 | 23 | 3213 | ||
NM_014971.1 | NP_055786.1 | 23 | 3225 | |||
XM_011532701.2 | XP_011531003.1 | 23 | 3198 | |||
GenBank mRNA | AK095190.1 | - | 24 | 3163 |