Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015123.2 | NP_055938.1 | 23 | 4531 | ||
XM_005264722.1 | XP_005264779.1 | 23 | 4208 | |||
XM_005264723.1 | XP_005264780.1 | 13 | 3690 | |||
XM_017005989.1 | XP_016861478.1 | 24 | 4407 | |||
XM_017005990.1 | XP_016861479.1 | 24 | 4300 | |||
XM_017005991.1 | XP_016861480.1 | 25 | 4460 | |||
XM_017005992.1 | XP_016861481.1 | 24 | 4240 | |||
XM_017005993.1 | XP_016861482.1 | 23 | 4139 | |||
XM_017005994.1 | XP_016861483.1 | 23 | 5981 | |||
XM_017005995.1 | XP_016861484.1 | 18 | 3817 | |||
XM_017005996.1 | XP_016861485.1 | 14 | 3519 | |||
XM_017005997.1 | XP_016861486.1 | 14 | 3782 | |||
GenBank mRNA | AB023230.1 | - | 23 | 4300 | ||
AK024086.1 | - | 7 | 2566 | |||
AL110231.1 | - | 1 | 574 | |||
BC028291.1 | - | 3 | 1880 | |||
BC110637.1 | - | 8 | 2752 |