Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBEAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015175.2 | NP_055990.1 | 38 | 6397 | ||
XM_006713072.3 | XP_006713135.1 | 37 | 6313 | |||
XM_011533533.2 | XP_011531835.1 | 38 | 6394 | |||
XM_017006010.1 | XP_016861499.1 | 38 | 6394 | |||
XM_017006011.1 | XP_016861500.1 | 38 | 6250 | |||
XM_017006012.1 | XP_016861501.1 | 37 | 6313 | |||
XM_017006013.1 | XP_016861502.1 | 38 | 6394 | |||
XM_017006014.1 | XP_016861503.1 | 37 | 6169 | |||
XM_017006015.1 | XP_016861504.1 | 36 | 6025 | |||
XM_017006016.1 | XP_016861505.1 | 33 | 6971 | |||
XM_017006017.1 | XP_016861506.1 | 15 | 2813 | |||
GenBank mRNA | AB011112.3 | - | 24 | 4081 | ||
AK131104.1 | - | 38 | 6237 | |||
AK301269.1 | - | 3 | 460 | |||
AY358455.1 | - | 8 | 1347 | |||
BC060874.1 | - | 6 | 875 |