Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001196.3 | NP_001187.1 | 5 | 613 | ||
NM_001244567.1 | NP_001231496.1 | 5 | 524 | |||
NM_001244569.1 | NP_001231498.1 | 4 | 402 | |||
NM_001244570.1 | NP_001231499.1 | 3 | 332 | |||
NM_001244572.1 | NP_001231501.1 | 3 | 650 | |||
NM_197966.2 | NP_932070.1 | 5 | 931 | |||
NM_197967.2 | NP_932071.1 | 4 | 543 | |||
XM_017028906.1 | XP_016884395.1 | 5 | 589 | |||
GenBank mRNA | AF042083.1 | - | 5 | 580 | ||
AF087891.1 | - | 5 | 578 | |||
AF250233.1 | - | 5 | 872 | |||
AF370380.1 | - | 2 | 1921 | |||
AK094795.1 | - | 5 | 872 | |||
AK313729.1 | - | 5 | 568 | |||
AY005151.1 | - | 6 | 625 | |||
BC009197.2 | - | 5 | 575 | 4025880 | ||
BC022072.1 | - | 5 | 613 | |||
BC033634.1 | - | 5 | 582 | 5538243 | ||
BC036364.1 | - | 5 | 906 | 4819949 | ||
BM561391.1 | - | 5 | 624 | |||
BM842561.1 | - | 4 | 543 | |||
BQ068464.1 | - | 5 | 591 | |||
CN277529.1 | - | 4 | 372 | |||
CR407603.1 | - | 3 | 440 | |||
CR456389.1 | - | 4 | 482 | |||
D89803.1 | - | 4 | 513 | |||
DA007047.1 | - | 3 | 326 | |||
DA158889.1 | - | 4 | 543 | |||
DB167767.1 | - | 3 | 312 | |||
EU447348.1 | - | 3 | 263 | |||
EU678292.1 | - | 5 | 1218 | |||
EU678293.1 | - | 3 | 243 | |||
EU678294.1 | - | 5 | 524 |