Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006721728.3 | XP_006721791.1 | 17 | 2600 | ||
XM_011524428.2 | XP_011522730.1 | 18 | 2377 | |||
XM_011524429.2 | XP_011522731.1 | 18 | 2534 | |||
XM_011524430.2 | XP_011522732.1 | 19 | 2500 | |||
XM_011524431.2 | XP_011522733.1 | 18 | 2432 | |||
XM_011524433.2 | XP_011522735.1 | 12 | 1790 | |||
XM_011524434.2 | XP_011522736.1 | 9 | 1237 | |||
XM_017024263.1 | XP_016879752.1 | 20 | 2594 | |||
XM_017024264.1 | XP_016879753.1 | 20 | 3579 | |||
XM_017024265.1 | XP_016879754.1 | 20 | 2788 | |||
XM_017024266.1 | XP_016879755.1 | 18 | 2549 | |||
XM_017024267.1 | XP_016879756.1 | 17 | 2401 | |||
XM_017024268.1 | XP_016879757.1 | 19 | 3485 | |||
XM_017024269.1 | XP_016879758.1 | 17 | 2674 |