Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EIF4G3 SIRNA Assays ›
Gene Symbol:
EIF4G3
Gene Name:
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases:
eIF-4G 3, eIF4G 3, eIF4GII
Chromosome Location:
Chr.1: 20806292 - 21177320 on Build GRCh38
UniGene ID:
Hs.467084
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
266746
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EIF4G3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_011542356.2
XP_011540658.1
1
400
XM_011542361.2
XP_011540663.1
1
400
XM_011542370.1
XP_011540672.1
1
400
XM_011542371.1
XP_011540673.1
1
400
XM_011542374.1
XP_011540676.1
1
400
XM_011542375.2
XP_011540677.1
1
400
XM_017002672.1
XP_016858161.1
1
400
XM_017002673.1
XP_016858162.1
1
400
XM_017002674.1
XP_016858163.1
1
400
XM_017002675.1
XP_016858164.1
1
400
XM_017002676.1
XP_016858165.1
1
400
XM_017002677.1
XP_016858166.1
1
400
XM_017002678.1
XP_016858167.1
1
400
XM_017002679.1
XP_016858168.1
1
400
XM_017002681.1
XP_016858170.1
1
400
XM_017002682.1
XP_016858171.1
1
400
XM_017002683.1
XP_016858172.1
1
400
XM_017002684.1
XP_016858173.1
1
400
XM_017002685.1
XP_016858174.1
1
400
XM_017002686.1
XP_016858175.1
1
400
XM_017002687.1
XP_016858176.1
1
400
XM_017002689.1
XP_016858178.1
1
400
XM_017002690.1
XP_016858179.1
1
400
XM_017002691.1
XP_016858180.1
1
400
XM_017002692.1
XP_016858181.1
1
400
XM_017002693.1
XP_016858182.1
1
400
XM_017002694.1
XP_016858183.1
1
400
XM_017002695.1
XP_016858184.1
1
400
XM_017002696.1
XP_016858185.1
1
400
XM_017002697.1
XP_016858186.1
1
400
XM_017002698.1
XP_016858187.1
1
400
XM_017002699.1
XP_016858188.1
1
400
XM_017002700.1
XP_016858189.1
1
400
XM_017002701.1
XP_016858190.1
1
400
XM_017002702.1
XP_016858191.1
1
400
XM_017002704.1
XP_016858193.1
1
400
XM_017002705.1
XP_016858194.1
1
400
XM_017002706.1
XP_016858195.1
1
400
XM_017002707.1
XP_016858196.1
1
400
XM_017002708.1
XP_016858197.1
1
400
XM_017002709.1
XP_016858198.1
1
400
XM_017002710.1
XP_016858199.1
1
400
XM_017002711.1
XP_016858200.1
1
400
XM_017002712.1
XP_016858201.1
1
400
Back To Top
More Information
Panther Classification:
Molecular Function -
translation factor
translation initiation factor
translational protein
Gene Ontology Categories:
Function(s)
Process(es)
translational initiation
regulation of translational initiation
viral process
RNA cap binding
translation initiation factor activity
translation factor activity, RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0