Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGGY SIRNA Assays ›
Gene Symbol:
FGGY
Gene Name:
FGGY carbohydrate kinase domain containing
Gene Aliases:
-
Chromosome Location:
Chr.1: 59296405 - 59762730 on Build GRCh38
UniGene ID:
Hs.444301
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
26695
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGGY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113411.1
NP_001106882.1
5
754
NM_001244714.1
NP_001231643.1
3
391
NM_018291.3
NP_060761.3
5
754
NR_103473.1
6
712
XM_011541730.1
XP_011540032.1
5
678
XM_011541731.1
XP_011540033.1
5
612
XM_011541732.1
XP_011540034.1
5
655
XM_011541733.1
XP_011540035.1
5
573
XM_011541735.1
XP_011540037.1
5
573
XM_011541736.2
XP_011540038.1
3
519
XM_017001643.1
XP_016857132.1
5
784
XM_017001644.1
XP_016857133.1
5
577
XM_017001645.1
XP_016857134.1
5
678
XM_017001646.1
XP_016857135.1
5
655
XM_017001647.1
XP_016857136.1
5
783
XM_017001648.1
XP_016857137.1
6
597
XM_017001649.1
XP_016857138.1
5
723
XM_017001650.1
XP_016857139.1
7
946
XM_017001651.1
XP_016857140.1
7
802
XM_017001652.1
XP_016857141.1
5
723
XM_017001653.1
XP_016857142.1
5
655
XM_017001654.1
XP_016857143.1
3
474
XM_017001655.1
XP_016857144.1
3
383
XM_017001656.1
XP_016857145.1
6
790
XM_017001657.1
XP_016857146.1
5
578
XM_017001658.1
XP_016857147.1
6
721
XM_017001659.1
XP_016857148.1
4
611
XM_017001660.1
XP_016857149.1
4
514
XM_017001661.1
XP_016857150.1
4
671
XM_017001662.1
XP_016857151.1
4
356
XM_017001663.1
XP_016857152.1
4
531
XM_017001664.1
XP_016857153.1
3
408
XM_017001665.1
XP_016857154.1
3
315
XM_017001666.1
XP_016857155.1
3
469
XM_017001667.1
XP_016857156.1
3
523
XM_017001668.1
XP_016857157.1
3
279
XM_017001669.1
XP_016857158.1
3
374
XM_017001670.1
XP_016857159.1
3
1147
XM_017001671.1
XP_016857160.1
3
275
XM_017001672.1
XP_016857161.1
4
611
XM_017001673.1
XP_016857162.1
4
668
XM_017001674.1
XP_016857163.1
4
527
XM_017001675.1
XP_016857164.1
4
611
XM_017001676.1
XP_016857165.1
4
671
XM_017001679.1
XP_016857168.1
5
723
GenBank mRNA
AK022237.1
-
4
611
AK054842.1
-
3
382
AK090568.1
-
6
712
AK315649.1
-
5
639
BC014947.1
-
5
639
4871664
BX647626.1
-
5
726
DA969344.1
-
4
539
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001013932, XM_008763856, XM_006238413, XM_006238410, BC090077.This siRNA also targets the following transcripts in Mouse: NM_001113412, NM_029347, BC031708, AK138439, AK085534.
Panther Classification:
Molecular Function -
carbohydrate kinase
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
phosphorylation
neuron cellular homeostasis
molecular_function
kinase activity
phosphotransferase activity, alcohol group as acceptor
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0