Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099334.2 | NP_001092804.1 | 9 | 1107 | ||
XM_017004074.1 | XP_016859563.1 | 9 | 1115 | |||
XM_017004075.1 | XP_016859564.1 | 9 | 1055 | |||
XM_017004076.1 | XP_016859565.1 | 9 | 1095 | |||
XM_017004077.1 | XP_016859566.1 | 8 | 936 | |||
GenBank mRNA | BC035737.1 | - | 9 | 1108 | 5730487 | |
DB541912.1 | - | 1 | 377 |