Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199898.1 | NP_001186827.1 | 9 | 1050 | ||
NM_001199899.1 | NP_001186828.1 | 8 | 849 | |||
NM_002611.4 | NP_002602.2 | 8 | 1012 | |||
GenBank mRNA | AK055119.1 | - | 9 | 1050 | ||
AK290522.1 | - | 8 | 911 | |||
AK293600.1 | - | 8 | 826 | |||
AK297089.1 | - | 8 | 799 | |||
BC005811.2 | - | 8 | 900 | 3831465 | ||
BC040478.1 | - | 8 | 955 | 4690182 | ||
BC063137.1 | - | 8 | 926 | 4272593 | ||
L42451.1 | - | 8 | 899 |