Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144960.2 | NP_001138432.1 | 3 | 711 | ||
NM_001315505.1 | NP_001302434.1 | 3 | 683 | |||
NM_001315506.1 | NP_001302435.1 | 3 | 711 | |||
XM_011512798.1 | XP_011511100.1 | 3 | 208 | |||
XM_011512799.1 | XP_011511101.1 | 3 | 442 | |||
XM_011512802.1 | XP_011511104.1 | 3 | 442 | |||
GenBank mRNA | AK098078.1 | - | 2 | 167 | ||
AK304359.1 | - | 3 | 442 | |||
BC171805.1 | - | 3 | 683 |