Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017006196.1 | XP_016861685.1 | 4 | 5588 | ||
XM_017006197.1 | XP_016861686.1 | 6 | 5840 | |||
XM_017006198.1 | XP_016861687.1 | 5 | 5653 | |||
XM_017006199.1 | XP_016861688.1 | 4 | 4650 | |||
XM_017006200.1 | XP_016861689.1 | 3 | 5508 | |||
XM_017006201.1 | XP_016861690.1 | 2 | 5357 | |||
GenBank mRNA | AK092973.1 | - | 1 | 3064 |