Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114726.2 | NP_001108198.2 | 2, 3 | 851 | ||
NM_001174164.1 | NP_001167635.1 | 3, 4 | 968 | |||
XM_005250342.3 | XP_005250399.1 | 2, 3 | 968 | |||
XM_005250343.3 | XP_005250400.1 | 2, 3 | 877 | |||
XM_006715985.3 | XP_006716048.1 | 2, 3 | 981 | |||
XM_017012205.1 | XP_016867694.1 | 3, 4 | 1787 | |||
XM_017012206.1 | XP_016867695.1 | 2, 3 | 1760 | |||
GenBank mRNA | BX327107.2 | - | 1, 2 | 133 |