Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018396.2 | NP_060866.2 | 3 | 384 | ||
GenBank mRNA | AK002212.1 | - | 2 | 257 | ||
AK299960.1 | - | 3 | 348 | |||
AK302775.1 | - | 3 | 365 | |||
BC064929.1 | - | 3 | 344 | |||
BC107586.1 | - | 2 | 268 | 6494734 | ||
BC121115.1 | - | 3 | 309 | |||
BC121116.1 | - | 3 | 309 |