Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MZT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001071775.2 | NP_001065243.1 | 3 | 1420 | ||
GenBank mRNA | AK025522.1 | - | 3 | 1293 | ||
BC047655.1 | - | 1 | 931 | |||
BC062361.1 | - | 3 | 1253 | |||
BC125183.1 | - | 3 | 1423 | 40122127 |