Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other F2 SIRNA Assays ›
          Gene Symbol:
          F2
          Gene Name:
          coagulation factor II, thrombin
          Gene Aliases:
          PT, RPRGL2, THPH1
          Chromosome Location:
          Chr.11: 46719166 - 46739508 on Build GRCh38
          UniGene ID:
          Hs.655207
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 2712

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: F2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000506.4 NP_000497.1 14 1923
          NM_001311257.1 NP_001298186.1 14 1875
          GenBank mRNA AJ972449.1 - 14 1884
          AK222775.1 - 14 1858
          AK222777.1 - 14 1858
          AK293326.1 - 14 1797
          AK303747.1 - 14 1837
          AK312965.1 - 14 1888
          AY344793.1 - 7 872
          AY344794.1 - 6 764
          BC051332.1 - 14 1864 6283420
          V00595.1 - 14 1834

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease serine protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          signal peptide processing
          proteolysis
          ER to Golgi vesicle-mediated transport
          acute-phase response
          cell surface receptor signaling pathway
          multicellular organism development
          blood coagulation
          blood coagulation, intrinsic pathway
          positive regulation of cell proliferation
          regulation of cell shape
          response to wounding
          regulation of gene expression
          negative regulation of platelet activation
          positive regulation of phosphatidylinositol 3-kinase signaling
          response to inactivity
          peptidyl-glutamic acid carboxylation
          platelet activation
          regulation of blood coagulation
          positive regulation of blood coagulation
          positive regulation of cell growth
          positive regulation of collagen biosynthetic process
          fibrinolysis
          cellular protein metabolic process
          negative regulation of proteolysis
          negative regulation of astrocyte differentiation
          leukocyte migration
          positive regulation of release of sequestered calcium ion into cytosol
          regulation of cytosolic calcium ion concentration
          negative regulation of fibrinolysis
          cellular response to mechanical stimulus
          positive regulation of phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of reactive oxygen species metabolic process
          serine-type endopeptidase activity
          receptor binding
          calcium ion binding
          protein binding
          growth factor activity
          thrombospondin receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0