Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018509.3 | NP_060979.2 | 6 | 847 | ||
GenBank mRNA | AF119857.1 | - | 3 | 205 | ||
AK025328.1 | - | 6 | 847 | |||
AK316593.1 | - | 4 | 349 | |||
BC017168.1 | - | 6 | 805 | 3907072 | ||
BC052279.1 | - | 6 | 765 | 4127952 |