Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GSDMB SIRNA Assays ›
Gene Symbol:
GSDMB
Gene Name:
gasdermin B
Gene Aliases:
GSDML, PP4052, PRO2521
Chromosome Location:
Chr.17: 39904595 - 39919377 on Build GRCh38
UniGene ID:
Hs.306777
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
27132
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GSDMB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042471.1
NP_001035936.1
2, 3
374
NM_001165958.1
NP_001159430.1
2, 3
374
NM_001165959.1
NP_001159431.1
2, 3
365
NM_018530.2
NP_061000.2
1, 2
467
XM_011525004.1
XP_011523306.1
2, 3
1770
XM_011525006.1
XP_011523308.1
2, 3
1761
XM_011525007.2
XP_011523309.1
2, 3
1092
XM_011525009.1
XP_011523311.1
2, 3
1450
XM_011525011.1
XP_011523313.1
2, 3
1101
XM_011525012.1
XP_011523314.1
2, 3
1441
XM_011525013.1
XP_011523315.1
3, 4
1159
XM_011525015.1
XP_011523317.1
3, 4
1150
XM_017024848.1
XP_016880337.1
2, 3
2093
XM_017024849.1
XP_016880338.1
3, 4
1802
XM_017024850.1
XP_016880339.1
1, 2
2304
XM_017024851.1
XP_016880340.1
1, 2
2304
XM_017024852.1
XP_016880341.1
1, 2
2304
XM_017024853.1
XP_016880342.1
1, 2
2304
XM_017024854.1
XP_016880343.1
1, 2
2304
GenBank mRNA
AF119884.1
-
1, 2
467
AK000409.1
-
2, 3
314
AK296607.1
-
2, 3
350
BC025682.1
-
1, 2
266
5205060
BP366854.1
-
1, 2
150
BX538068.1
-
1, 2
38
BX647700.1
-
2, 3
3660
EU526753.1
-
1, 2
243
EU526754.1
-
1, 2
243
EU526755.1
-
1, 2
243
EU526756.1
-
1, 2
243
Back To Top
More Information
Gene Ontology Categories:
Function(s)
programmed cell death
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mfv9s:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0