Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
企業服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MICAL3 SIRNA Assays ›
Gene Symbol:
MICAL3
Gene Name:
microtubule associated monooxygenase, calponin and LIM domain containing 3
Gene Aliases:
MICAL-3
Chromosome Location:
Chr.22: 17787649 - 18024559 on Build GRCh38
UniGene ID:
Hs.528024
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
272693
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MICAL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015241.2
NP_056056.2
32
7065
XM_005261262.3
XP_005261319.1
32
6895
XM_011546137.2
XP_011544439.1
40
7516
XM_011546139.2
XP_011544441.1
39
7465
XM_011546142.2
XP_011544444.1
38
7414
XM_017028865.1
XP_016884354.1
40
8034
XM_017028866.1
XP_016884355.1
40
7840
XM_017028867.1
XP_016884356.1
40
8957
XM_017028868.1
XP_016884357.1
40
7686
XM_017028869.1
XP_016884358.1
40
7431
XM_017028870.1
XP_016884359.1
40
7432
XM_017028871.1
XP_016884360.1
40
7513
XM_017028872.1
XP_016884361.1
39
7465
XM_017028873.1
XP_016884362.1
39
7453
XM_017028874.1
XP_016884363.1
39
7432
XM_017028875.1
XP_016884364.1
39
7408
XM_017028876.1
XP_016884365.1
39
7378
XM_017028877.1
XP_016884366.1
38
7315
XM_017028878.1
XP_016884367.1
37
7201
XM_017028879.1
XP_016884368.1
35
7060
XM_017028880.1
XP_016884369.1
35
7383
XM_017028881.1
XP_016884370.1
35
7230
XM_017028882.1
XP_016884371.1
34
6997
XM_017028883.1
XP_016884372.1
39
7179
XM_017028884.1
XP_016884373.1
36
7509
XM_017028885.1
XP_016884374.1
20
5101
XM_017028886.1
XP_016884375.1
20
4949
GenBank mRNA
AB020626.1
-
12
3673
AK126925.1
-
5
1951
BC006562.2
-
7
1839
BC047315.1
-
1
136
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
cytoskeleton organization
actin filament depolymerization
oxidation-reduction process
actin binding
zinc ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
Rab GTPase binding
FAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-5btgp:80/100.66.77.235:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0