Search
Search

FAM114A2| Gene Symbol: FAM114A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317993.1 | NP_001304922.1 | 5 | 467 | ||
| NM_001317994.1 | NP_001304923.1 | 4 | 984 | |||
| NM_001317995.1 | NP_001304924.1 | 2, 3 | 210 | |||
| NM_018691.3 | NP_061161.2 | 4 | 420 | |||
| XM_005268360.4 | XP_005268417.1 | 5 | 545 | |||
| XM_005268361.4 | XP_005268418.1 | 4 | 448 | |||
| XM_011537540.2 | XP_011535842.1 | 4 | 774 | |||
| XM_011537541.2 | XP_011535843.1 | 4 | 1356 | |||
| XM_011537542.2 | XP_011535844.1 | 4 | 645 | |||
| XM_017008967.1 | XP_016864456.1 | 4 | 439 | |||
| XM_017008968.1 | XP_016864457.1 | 4 | 461 | |||
| GenBank mRNA | AF159700.1 | - | 4 | 913 | ||
| AK024664.1 | - | 4 | 385 | |||
| AK225821.1 | - | 4 | 403 | |||
| AK298943.1 | - | 2, 3 | 179 | |||
| AK313331.1 | - | 4 | 392 | |||
| BC011414.1 | - | 5 | 402 | 3855873 | ||
| BC050701.1 | - | 4 | 381 | |||
| DA351656.1 | - | 4 | 420 | |||
| DB022316.1 | - | 5 | 530 | |||