Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100917.1 | NP_001094387.1 | 5, 6 | 445 | ||
XM_005268669.4 | XP_005268726.1 | 5, 6 | 495 | |||
XM_017018862.1 | XP_016874351.1 | 4, 5 | 383 | |||
XM_017018863.1 | XP_016874352.1 | 4, 5 | 383 | |||
XM_017018864.1 | XP_016874353.1 | 4, 5 | 383 | |||
GenBank mRNA | AK126325.1 | - | 5, 6 | 441 |