Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT8 SIRNA Assays ›
Gene Symbol:
SEPT8
Gene Name:
septin 8
Gene Aliases:
SEP2
Chromosome Location:
Chr.5: 132750817 - 132778216 on Build GRCh38
UniGene ID:
Hs.522057
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
273361
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098811.1
NP_001092281.1
6, 7
1038
NM_001098812.1
NP_001092282.1
6, 7
1038
NM_001098813.1
NP_001092283.1
6, 7
964
NM_001300798.1
NP_001287727.1
6, 7
1032
NM_001300799.1
NP_001287728.1
6, 7
1032
NM_015146.1
NP_055961.1
6, 7
1038
XM_005271927.2
XP_005271984.1
6, 7
1040
XM_005271928.2
XP_005271985.1
6, 7
1034
XM_005271931.3
XP_005271988.1
7, 8
1121
XM_011543288.1
XP_011541590.1
6, 7
1060
XM_017009254.1
XP_016864743.1
6, 7
1034
XM_017009255.1
XP_016864744.1
6, 7
1107
XM_017009256.1
XP_016864745.1
6, 7
1101
XM_017009257.1
XP_016864746.1
6, 7
1054
XM_017009258.1
XP_016864747.1
6, 7
1107
XM_017009259.1
XP_016864748.1
6, 7
1101
XM_017009260.1
XP_016864749.1
6, 7
1054
XM_017009261.1
XP_016864750.1
6, 7
1101
GenBank mRNA
AF179995.1
-
6, 7
1038
AF440761.1
-
6, 7
1038
AF440762.1
-
6, 7
1038
AF440763.1
-
6, 7
873
AK057797.1
-
6, 7
896
BC001329.1
-
3, 4
437
BC140759.1
-
6, 7
854
9021627
BC171779.1
-
6, 7
854
9054433
D86957.1
-
6, 7
1038
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_033144, NM_001252333, NM_001252332, AF179996, BC037707, and others.
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Process(es)
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-6dgdk:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0