Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015164.2 | NP_055979.2 | 10 | 2047 | ||
XM_005245790.3 | XP_005245847.1 | 9 | 2182 | |||
XM_005245791.4 | XP_005245848.1 | 5 | 1315 | |||
XM_017000757.1 | XP_016856246.1 | 10 | 1954 | |||
XM_017000758.1 | XP_016856247.1 | 9 | 1894 | |||
GenBank mRNA | AB020649.1 | - | 10 | 1823 | ||
BC008002.1 | - | 2 | 84 | |||
BC016488.1 | - | 2 | 729 | |||
BC030545.1 | - | 1 | 27 | |||
BC040441.1 | - | 10 | 2050 | |||
BC042103.1 | - | 2 | 377 | |||
BC068599.1 | - | 2 | 402 |