Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015295.2 | NP_056110.2 | 5 | 723 | ||
XM_011525642.1 | XP_011523944.1 | 5 | 723 | |||
XM_011525643.2 | XP_011523945.1 | 5 | 723 | |||
XM_011525644.2 | XP_011523946.1 | 4 | 631 | |||
XM_017025684.1 | XP_016881173.1 | 6 | 901 | |||
GenBank mRNA | BC006008.1 | - | 5 | 723 |