Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257158.1 | NP_001244087.1 | 5 | 510 | ||
NM_001257159.1 | NP_001244088.1 | 4 | 462 | |||
NM_018718.2 | NP_061188.1 | 5 | 510 | |||
NR_046443.1 | 5 | 510 | ||||
XM_011516709.2 | XP_011515011.1 | 6 | 471 | |||
XM_011516710.2 | XP_011515012.1 | 4 | 286 | |||
GenBank mRNA | AF429308.1 | - | 5 | 356 | ||
AJ278890.1 | - | 5 | 313 | |||
AK026098.1 | - | 5 | 310 | |||
AK298618.1 | - | 4 | 278 | |||
AK311382.1 | - | 5 | 327 | |||
AK314676.1 | - | 5 | 289 | |||
BC018789.1 | - | 5 | 292 | |||
BC056162.1 | - | 5 | 283 | 6142659 | ||
DB069500.1 | - | 5 | 512 |