Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CLU SIRNA Assays ›
          Gene Symbol:
          CLU
          Gene Name:
          clusterin
          Gene Aliases:
          AAG4, APO-J, APOJ, CLI, CLU1, CLU2, KUB1, NA1/NA2, SGP-2, SGP2, SP-40, TRPM-2, TRPM2
          Chromosome Location:
          Chr.8: 27596917 - 27616717 on Build GRCh38
          UniGene ID:
          Hs.436657
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 2745

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CLU
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001831.3 NP_001822.3 8 1468
          NR_038335.1 8 1603
          NR_045494.1 8 1462
          XM_006716284.2 XP_006716347.1 8 3374
          GenBank mRNA AK093399.1 - 9 1218
          AK301324.1 - 8 1072
          AK313870.1 - 8 1357
          AY513288.1 - 8 1347
          BC010514.1 - 8 1350 4150452
          BC013221.1 - 7 1283
          BC019588.2 - 7 1318 4915444
          BC029972.1 - 7 2494
          BX648414.1 - 7 2068
          J02908.1 - 6 1200
          M25915.1 - 8 1477
          M64722.1 - 8 1334
          M74816.1 - 6 1104
          X14723.1 - 7 1329

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          microglial cell activation
          release of cytochrome c from mitochondria
          platelet degranulation
          lipid metabolic process
          complement activation
          complement activation, classical pathway
          aging
          positive regulation of cell proliferation
          response to light stimulus
          response to wounding
          response to virus
          protein import
          endocrine pancreas development
          central nervous system myelin maintenance
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of protein homooligomerization
          positive regulation of tumor necrosis factor production
          response to potassium ion
          reverse cholesterol transport
          estrous cycle
          innate immune response
          positive regulation of nitric oxide biosynthetic process
          positive regulation of cell differentiation
          neuron projection morphogenesis
          protein stabilization
          positive regulation of NF-kappaB transcription factor activity
          chaperone-mediated protein complex assembly
          response to misfolded protein
          chaperone-mediated protein folding
          microglial cell proliferation
          cellular response to growth factor stimulus
          regulation of beta-amyloid clearance
          regulation of neuron death
          positive regulation of neuron death
          positive regulation of beta-amyloid formation
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of beta-amyloid formation
          regulation of neuronal signal transduction
          positive regulation of tau-protein kinase activity
          positive regulation of neurofibrillary tangle assembly
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          protein binding
          ATPase activity
          ubiquitin protein ligase binding
          protein N-terminus binding
          chaperone binding
          misfolded protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0