Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018897.2 | NP_061720.2 | 4 | 277 | ||
XM_011511487.2 | XP_011509789.1 | 5 | 653 | |||
XM_011511488.2 | XP_011509790.1 | 4 | 1447 | |||
XM_011511489.2 | XP_011509791.1 | 3 | 239 | |||
XM_011511490.2 | XP_011509792.1 | 4 | 1447 | |||
XM_011511491.2 | XP_011509793.1 | 4 | 1449 | |||
XM_011511492.2 | XP_011509794.1 | 4 | 1449 | |||
XM_011511493.2 | XP_011509795.1 | 4 | 1449 | |||
XM_011511494.2 | XP_011509796.1 | 4 | 1448 | |||
XM_011511495.2 | XP_011509797.1 | 4 | 1448 | |||
XM_017004504.1 | XP_016859993.1 | 4 | 1446 | |||
GenBank mRNA | AB023161.2 | - | 4 | 295 | ||
AF327442.1 | - | 4 | 272 | |||
BP199399.1 | - | 4 | 277 |