Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FXR1 SIRNA Assays ›
          Gene Symbol:
          FXR1
          Gene Name:
          FMR1 autosomal homolog 1
          Gene Aliases:
          FXR1P
          Chromosome Location:
          Chr.3: 180912446 - 180982753 on Build GRCh38
          UniGene ID:
          Hs.478407
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 2754

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FXR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001013438.2 NP_001013456.1 3, 4 446
          NM_001013439.2 NP_001013457.1 4, 5 790
          NM_005087.3 NP_005078.2 3, 4 446
          XM_005247813.3 XP_005247870.2 3, 4 221
          XM_005247814.3 XP_005247871.2 3, 4 221
          XM_005247815.3 XP_005247872.2 3, 4 221
          XM_005247816.3 XP_005247873.2 3, 4 221
          XM_006713775.3 XP_006713838.1 4, 5 564
          XM_017007298.1 XP_016862787.1 4, 5 564
          XM_017007299.1 XP_016862788.1 4, 5 564
          XM_017007300.1 XP_016862789.1 4, 5 564
          XM_017007301.1 XP_016862790.1 4, 5 564
          GenBank mRNA AK225334.1 - 3, 4 222
          AK292633.1 - 3, 4 222
          AK294603.1 - 2, 3 168
          AK302197.1 - 3, 4 225
          AY341428.1 - 3, 4 497
          BC028983.1 - 3, 4 216 5122574
          BX956500.1 - 3, 4 232
          DC352767.1 - 3, 4 446
          KR259877.1 - 3, 4 206
          KR259878.1 - 3, 4 206
          KR259879.1 - 3, 4 206
          KR259880.1 - 4, 5 550
          U25165.1 - 3, 4 218

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_008760951, XM_008760952, BC083807.
          This siRNA also targets the following transcripts in Mouse: NM_001113188, NM_008053, NM_001113189, BC019139, X90875, and others.

          Panther Classification:

          Molecular Function -

          translation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          muscle organ development
          negative regulation of translation
          cell differentiation
          positive regulation of gene silencing by miRNA
          G-quadruplex RNA binding
          RNA binding
          mRNA binding
          mRNA 3'-UTR binding
          protein binding
          RNA strand annealing activity
          protein homodimerization activity
          poly(A) RNA binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gbx7x:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0