Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD34C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146341.1 | NP_001139813.1 | 2 | 2024 | ||
GenBank mRNA | AK127037.1 | - | 3 | 1709 |