Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MOV10L1 SIRNA Assays ›
          Gene Symbol:
          MOV10L1
          Gene Name:
          Mov10 RISC complex RNA helicase like 1
          Gene Aliases:
          CHAMP, DJ402G11.8
          Chromosome Location:
          Chr.22: 50090006 - 50161705 on Build GRCh38
          UniGene ID:
          Hs.62880
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 27766

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MOV10L1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164104.1 NP_001157576.1 2, 3 372
          NM_001164105.1 NP_001157577.1 2, 3 449
          NM_018995.2 NP_061868.1 2, 3 372
          XM_005261923.3 XP_005261980.1 2, 3 650
          XM_011530696.1 XP_011528998.1 2, 3 372
          XM_011530697.1 XP_011528999.1 2, 3 650
          XM_011530698.1 XP_011529000.1 2, 3 372
          XM_011530699.1 XP_011529001.1 2, 3 334
          XM_011530700.2 XP_011529002.1 2, 3 717
          XM_011530701.1 XP_011529003.1 2, 3 372
          XM_011530704.1 XP_011529006.1 2, 3 372
          XM_017028833.1 XP_016884322.1 2, 3 372
          XM_017028834.1 XP_016884323.1 2, 3 372
          XM_017028837.1 XP_016884326.1 2, 3 372
          GenBank mRNA AF285604.1 - 2, 3 316
          AK302401.1 - 2, 3 449
          BC029552.1 - 2, 3 652
          BC042828.1 - 2, 3 652
          BC140944.1 - 3, 4 373 9056756
          BC150137.1 - 2, 3 314 40015813
          BC152539.1 - 2, 3 314 40009193
          CR456466.1 - 2, 3 295
          DB451595.2 - 2, 3 372

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: BC125262.

          Gene Ontology Categories:

          Function(s) Process(es)

          male meiosis I
          multicellular organism development
          germ cell development
          spermatogenesis
          piRNA metabolic process
          DNA methylation involved in gamete generation
          magnesium ion binding
          RNA binding
          ATP-dependent RNA helicase activity
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0