Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142549.1 | NP_001136021.1 | 3 | 214 | ||
NM_001166533.1 | NP_001160005.1 | 3 | 214 | |||
NM_024415.2 | NP_077726.1 | 3 | 214 | |||
XM_011543495.2 | XP_011541797.1 | 3 | 237 | |||
XM_011543497.2 | XP_011541799.1 | 3 | 237 | |||
GenBank mRNA | AF262962.1 | - | 2 | 132 | ||
AK292417.1 | - | 3 | 189 | |||
AK308300.1 | - | 3 | 191 | |||
AY004154.1 | - | 2 | 128 | |||
BC047455.1 | - | 3 | 152 | 5298807 | ||
BC088362.1 | - | 3 | 173 | 30386875 | ||
BI559479.1 | - | 3 | 176 | |||
DB457419.1 | - | 3 | 214 |