Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRAMD1B SIRNA Assays ›
Gene Symbol:
GRAMD1B
Gene Name:
GRAM domain containing 1B
Gene Aliases:
-
Chromosome Location:
Chr.11: 123358299 - 123627771 on Build GRCh38
UniGene ID:
Hs.144725
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
278815
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRAMD1B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286563.1
NP_001273492.1
7, 8
1082
NM_001286564.1
NP_001273493.1
6, 7
791
NM_020716.2
NP_065767.1
6, 7
1061
XM_005271619.4
XP_005271676.1
6, 7
1061
XM_005271621.4
XP_005271678.1
7, 8
5448
XM_005271623.4
XP_005271680.1
6, 7
826
XM_005271624.4
XP_005271681.2
6, 7
741
XM_006718892.3
XP_006718955.1
7, 8
978
XM_011542926.2
XP_011541228.1
7, 8
975
XM_011542929.2
XP_011541231.1
6, 7
951
XM_011542930.2
XP_011541232.1
6, 7
1592
XM_011542931.2
XP_011541233.1
6, 7
1506
XM_011542933.1
XP_011541235.1
4, 5
1203
XM_017018034.1
XP_016873523.1
8, 9
1046
XM_017018035.1
XP_016873524.1
8, 9
1043
XM_017018036.1
XP_016873525.1
7, 8
1513
XM_017018037.1
XP_016873526.1
8, 9
1812
XM_017018038.1
XP_016873527.1
8, 9
1046
XM_017018039.1
XP_016873528.1
7, 8
1104
XM_017018040.1
XP_016873529.1
6, 7
900
XM_017018041.1
XP_016873530.1
7, 8
1104
XM_017018042.1
XP_016873531.1
7, 8
1104
XM_017018043.1
XP_016873532.1
6, 7
737
XM_017018044.1
XP_016873533.1
6, 7
1061
XM_017018045.1
XP_016873534.1
7, 8
5445
XM_017018046.1
XP_016873535.1
7, 8
1550
XM_017018047.1
XP_016873536.1
7, 8
1547
XM_017018048.1
XP_016873537.1
6, 7
1503
XM_017018049.1
XP_016873538.1
7, 8
1544
XM_017018050.1
XP_016873539.1
6, 7
820
XM_017018051.1
XP_016873540.1
4, 5
424
GenBank mRNA
AB033027.1
-
6, 7
724
AK127457.1
-
7, 8
1082
AK297606.1
-
6, 7
791
AK307795.1
-
6, 7
901
BC105741.1
-
6, 7
875
40026783
BC105931.1
-
6, 7
875
40026781
BC105932.1
-
6, 7
875
40026782
BC107480.1
-
6, 7
875
40026784
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-gzxth:80/10.239.166.29:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0