Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080423.3 | NP_001073892.3 | 24 | 5750 | ||
GenBank mRNA | AB051506.2 | - | 24 | 5736 |