Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008778.1 | NP_001008778.1 | 3 | 224 | ||
XM_017017705.1 | XP_016873194.1 | 2 | 618 | |||
XM_017017706.1 | XP_016873195.1 | 2 | 618 | |||
XM_017017707.1 | XP_016873196.1 | 2 | 618 | |||
XM_017017708.1 | XP_016873197.1 | 2 | 618 | |||
XM_017017709.1 | XP_016873198.1 | 2 | 618 | |||
XM_017017710.1 | XP_016873199.1 | 2 | 618 | |||
GenBank mRNA | AY820305.1 | - | 3 | 224 | ||
BC137244.1 | - | 3 | 306 | 9021241 | ||
BC137246.1 | - | 3 | 306 | 9021243 | ||
BC144548.1 | - | 3 | 306 | 9053075 |