Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054027.4 | NP_473368.1 | 2 | 465 | ||
XM_011514067.1 | XP_011512369.1 | 2 | 428 | |||
XM_017009644.1 | XP_016865133.1 | 2 | 65 | |||
GenBank mRNA | AB046801.1 | - | 2 | 295 | ||
AF274753.1 | - | 2 | 202 | |||
AK222631.1 | - | 163 | ||||
AK315012.1 | - | 2 | 163 | |||
AY358503.1 | - | 2 | 216 | |||
BC009835.2 | - | 2 | 390 | 3927236 | ||
BC014526.2 | - | 2 | 190 | 3837372 | ||
BU193060.1 | - | 2 | 457 | |||
DA126651.1 | - | 2 | 428 |