Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPANXN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009613.2 | NP_001009613.1 | 2 | 303 | ||
XM_017029543.1 | XP_016885032.1 | 2 | 293 | |||
XM_017029544.1 | XP_016885033.1 | 2 | 290 | |||
GenBank mRNA | BC130503.1 | - | 2 | 303 | 40146534 | |
BC130505.1 | - | 2 | 303 | 40146536 | ||
DQ336126.1 | - | 2 | 206 | |||
DQ336127.1 | - | 2 | 206 |