Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF4 SIRNA Assays ›
Gene Symbol:
CELF4
Gene Name:
CUGBP, Elav-like family member 4
Gene Aliases:
BRUNOL4, CELF-4
Chromosome Location:
Chr.18: 37243040 - 37566052 on Build GRCh38
UniGene ID:
Hs.435976
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28023
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025087.1
NP_001020258.1
1
629
NM_001025088.1
NP_001020259.1
1
629
NM_001025089.1
NP_001020260.1
1
629
NM_020180.3
NP_064565.1
1
629
XM_005258308.3
XP_005258365.1
1
641
XM_011526079.2
XP_011524381.1
1
643
XM_011526081.2
XP_011524383.1
1
480
XM_011526083.2
XP_011524385.1
1
643
XM_011526084.2
XP_011524386.1
1
643
XM_011526085.2
XP_011524387.1
1
644
XM_011526086.2
XP_011524388.1
1
642
XM_011526087.2
XP_011524389.1
1
644
XM_011526090.2
XP_011524392.1
1
641
XM_011526091.2
XP_011524393.1
1
641
XM_011526092.2
XP_011524394.1
1
641
XM_017025840.1
XP_016881329.1
1
506
XM_017025841.1
XP_016881330.1
1
640
XM_017025842.1
XP_016881331.1
1
642
XM_017025843.1
XP_016881332.1
1
642
XM_017025844.1
XP_016881333.1
1
642
XM_017025845.1
XP_016881334.1
1
642
XM_017025846.1
XP_016881335.1
1
640
XM_017025847.1
XP_016881336.1
1
640
XM_017025848.1
XP_016881337.1
1
641
XM_017025849.1
XP_016881338.1
1
641
XM_017025850.1
XP_016881339.1
1
506
XM_017025851.1
XP_016881340.1
1
506
XM_017025852.1
XP_016881341.1
1
506
XM_017025853.1
XP_016881342.1
1
642
XM_017025854.1
XP_016881343.1
1
642
XM_017025855.1
XP_016881344.1
1
642
XM_017025856.1
XP_016881345.1
1
642
XM_017025857.1
XP_016881346.1
1
643
XM_017025858.1
XP_016881347.1
1
480
XM_017025859.1
XP_016881348.1
1
480
XM_017025860.1
XP_016881349.1
1
641
XM_017025861.1
XP_016881350.1
1
506
XM_017025862.1
XP_016881351.1
1
506
XM_017025863.1
XP_016881352.1
1
488
XM_017025864.1
XP_016881353.1
1
480
XM_017025865.1
XP_016881354.1
1
506
GenBank mRNA
AB209774.1
-
1
423
AF248650.1
-
1
368
AF329265.1
-
1
233
AK074596.1
-
1
361
AK295009.1
-
1
243
BC001946.1
-
1
390
2820541
BC004167.2
-
1
390
2820541
BC045711.1
-
1
316
5261155
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
alternative mRNA splicing, via spliceosome
regulation of alternative mRNA splicing, via spliceosome
mRNA splice site selection
germ cell development
embryo development
negative regulation of translation
negative regulation of mRNA splicing, via spliceosome
positive regulation of mRNA splicing, via spliceosome
negative regulation of excitatory postsynaptic potential
nucleotide binding
translation repressor activity, nucleic acid binding
mRNA binding
BRE binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0