Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@61f80164
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Contact Us
Sign In
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
Gene Symbol:
PARD3
Gene Name:
par-3 family cell polarity regulator
Gene Aliases:
ASIP, Baz, PAR3, PAR3alpha, PARD-3, PARD3A, PPP1R118, SE2-5L16, SE2-5LT1, SE2-5T2
Chromosome Location:
Chr.10: 34109560 - 34815325 on Build GRCh38
UniGene ID:
Hs.131489
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28053
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price (USD)
233.00
Your Price (USD)
Online offer (USD):
233.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PARD3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184785.1
NP_001171714.1
2
507
NM_001184786.1
NP_001171715.1
2
507
NM_001184787.1
NP_001171716.1
2
507
NM_001184788.1
NP_001171717.1
2
507
NM_001184789.1
NP_001171718.1
2
507
NM_001184790.1
NP_001171719.1
2
507
NM_001184791.1
NP_001171720.1
2
507
NM_001184792.1
NP_001171721.1
2
507
NM_001184793.1
NP_001171722.1
2
507
NM_001184794.1
NP_001171723.1
2
507
NM_019619.3
NP_062565.2
2
507
XM_005252528.4
XP_005252585.1
2
498
XM_005252530.4
XP_005252587.1
2
498
XM_005252531.4
XP_005252588.1
2
498
XM_005252532.4
XP_005252589.1
2
284
XM_005252534.4
XP_005252591.1
2
498
XM_005252535.4
XP_005252592.1
2
498
XM_005252536.4
XP_005252593.1
2
498
XM_011519569.2
XP_011517871.1
2
498
XM_011519570.2
XP_011517872.1
2
498
XM_011519571.2
XP_011517873.1
2
498
XM_011519572.2
XP_011517874.1
2
498
XM_011519573.2
XP_011517875.1
2
498
XM_011519576.2
XP_011517878.1
2
285
XM_011519577.2
XP_011517879.1
2
498
XM_011519578.2
XP_011517880.1
2
502
XM_011519581.2
XP_011517883.1
2
498
XM_011519583.2
XP_011517885.1
2
498
XM_011519584.2
XP_011517886.1
2
498
XM_017016430.1
XP_016871919.1
2
498
GenBank mRNA
AB073671.1
-
2
177
AF196185.1
-
2
464
AF196186.1
-
2
464
AF252293.1
-
2
177
AF332592.1
-
2
177
AF332593.1
-
2
177
AF454058.1
-
2
521
AF467002.1
-
2
177
AF467003.1
-
2
177
AF467004.1
-
2
177
AF467005.1
-
2
177
AF467006.1
-
2
177
BC071566.1
-
2
454
30344732
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-3-prod-green:8080/10.253.229.154:8080.
git-commit: 0f6c802f6da5e59cd1b986a4dc444b4bcf3ac46d
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.15.0-2021.01.02-1.0