Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC298111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024248.1 | NP_001019419.1 | 286 |
Gene Symbol: LOC103690101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763753.1 | XP_008761975.1 | 3 | 286 |