Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam89b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015013.1 | NP_001015013.1 | 1, 2 | 410 | ||
GenBank mRNA | BC093377.1 | - | 1, 2 | 410 | 7388027 |