Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HNRNPC SIRNA Assays ›
          Gene Symbol:
          Hnrnpc LOC100911576
          Gene Name:
          heterogeneous nuclear ribonucleoprotein C
          heterogeneous nuclear ribonucleoproteins C1/C2-like
          Gene Aliases:
          Hnrpc, hnRNP C
          -
          Chromosome Location:
          Chr.15: 28486605 - 28517353 on Build Rnor_6.0
          Chromosome Location:
          Chr.14: 88543630 - 88574207 on Build Rnor_6.0
          UniGene ID:
          Rn.101929
          Rn.101929
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 280764

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hnrnpc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277600.1 NP_001264529.1 7 821
          NM_001277601.1 NP_001264530.1 7 782
          NM_001277602.1 NP_001264531.1 6 756
          XM_006251858.2 XP_006251920.1 6 1289
          XM_006251859.2 XP_006251921.1 6 782
          XM_006251861.2 XP_006251923.1 6 1249
          XM_006251862.2 XP_006251924.1 8 933
          XM_006251863.2 XP_006251925.1 6 743
          XM_006251864.2 XP_006251926.1 8 838
          XM_006251865.2 XP_006251927.1 6 1249
          XM_006251866.1 XP_006251928.1 7 769
          XM_008770508.1 XP_008768730.1 7 1316
          XM_008770509.1 XP_008768731.1 7 2384
          XM_008770510.1 XP_008768732.1 7 816
          XM_008770511.1 XP_008768733.1 7 816
          XM_008770512.1 XP_008768734.1 7 769
          GenBank mRNA BC097346.1 - 6 700 7442283
          FQ214011.1 - 5 390
          FQ222540.1 - 6 432
          FQ227266.1 - 5 432
          Gene Symbol: LOC100911576
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271241.1 NP_001258170.1 7 747
          XM_003751394.2 XP_003751442.1 7 733
          XM_003751395.2 XP_003751443.1 7 772
          XM_006251449.2 XP_006251511.1 7 2383
          XM_006251450.2 XP_006251512.1 6 1287
          XM_006251451.1 XP_006251513.1 6 746
          XM_006251452.2 XP_006251514.1 7 772
          XM_006251453.1 XP_006251515.1 8 897
          XM_006251454.1 XP_006251516.1 8 802
          XM_006251455.2 XP_006251517.1 6 1247
          XM_006251456.1 XP_006251518.1 6 707
          XM_006251457.1 XP_006251519.1 6 707
          XM_006251458.2 XP_006251520.1 7 733
          XM_008770242.1 XP_008768464.1 7 1280
          XM_008770243.1 XP_008768465.1 7 772
          GenBank mRNA AY724499.1 - 6 1175
          FQ214198.1 - 5 432
          FQ215781.1 - 5 471
          FQ227893.1 - 5 432
          FQ228082.1 - 5 471

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001170983, NM_001170984, NM_001170982, NM_001170981, NM_016884, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          mRNA processing
          RNA splicing
          ATP-dependent chromatin remodeling
          3'-UTR-mediated mRNA stabilization
          nucleotide binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          nucleic acid binding
          RNA binding
          mRNA binding
          mRNA 3'-UTR binding
          poly(U) RNA binding
          nucleosomal DNA binding
          identical protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0