Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HNRNPC SIRNA Assays ›
          Gene Symbol:
          Hnrnpc LOC100911576
          Gene Name:
          heterogeneous nuclear ribonucleoprotein C
          heterogeneous nuclear ribonucleoproteins C1/C2-like
          Gene Aliases:
          Hnrpc, hnRNP C
          -
          Chromosome Location:
          Chr.15: 28486605 - 28517353 on Build Rnor_6.0
          Chromosome Location:
          Chr.14: 88543630 - 88574207 on Build Rnor_6.0
          UniGene ID:
          Rn.101929
          Rn.101929
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 280765

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hnrnpc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277600.1 NP_001264529.1 5, 6 612
          NM_001277601.1 NP_001264530.1 5, 6 573
          NM_001277602.1 NP_001264531.1 4, 5 547
          XM_006251858.2 XP_006251920.1 4, 5 1080
          XM_006251859.2 XP_006251921.1 4, 5 573
          XM_006251861.2 XP_006251923.1 4, 5 1040
          XM_006251862.2 XP_006251924.1 6, 7 724
          XM_006251863.2 XP_006251925.1 4, 5 534
          XM_006251864.2 XP_006251926.1 6, 7 629
          XM_006251865.2 XP_006251927.1 4, 5 1040
          XM_006251866.1 XP_006251928.1 5, 6 560
          XM_008770508.1 XP_008768730.1 5, 6 1107
          XM_008770509.1 XP_008768731.1 5, 6 2175
          XM_008770510.1 XP_008768732.1 5, 6 607
          XM_008770511.1 XP_008768733.1 5, 6 607
          XM_008770512.1 XP_008768734.1 5, 6 560
          GenBank mRNA BC097346.1 - 4, 5 497 7442283
          CB582273.1 - 5, 6 562
          FQ214011.1 - 3, 4 184
          FQ227266.1 - 3, 4 223
          Gene Symbol: LOC100911576
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271241.1 NP_001258170.1 5, 6 538
          XM_003751394.2 XP_003751442.1 5, 6 524
          XM_003751395.2 XP_003751443.1 5, 6 563
          XM_006251449.2 XP_006251511.1 5, 6 2174
          XM_006251450.2 XP_006251512.1 4, 5 1078
          XM_006251451.1 XP_006251513.1 4, 5 537
          XM_006251452.2 XP_006251514.1 5, 6 563
          XM_006251453.1 XP_006251515.1 6, 7 688
          XM_006251454.1 XP_006251516.1 6, 7 593
          XM_006251455.2 XP_006251517.1 4, 5 1038
          XM_006251456.1 XP_006251518.1 4, 5 498
          XM_006251457.1 XP_006251519.1 4, 5 498
          XM_006251458.2 XP_006251520.1 5, 6 524
          XM_008770242.1 XP_008768464.1 5, 6 1071
          XM_008770243.1 XP_008768465.1 5, 6 563
          GenBank mRNA AY724499.1 - 4, 5 966
          FQ214198.1 - 3, 4 223
          FQ215781.1 - 3, 4 262
          FQ227893.1 - 3, 4 223
          FQ228082.1 - 3, 4 262

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          mRNA processing
          RNA splicing
          ATP-dependent chromatin remodeling
          3'-UTR-mediated mRNA stabilization
          nucleotide binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          nucleic acid binding
          RNA binding
          mRNA binding
          mRNA 3'-UTR binding
          poly(U) RNA binding
          nucleosomal DNA binding
          identical protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7wptj:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0