Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256279.1 | NP_001243208.1 | 5 | 2396 | ||
NM_001256280.1 | NP_001243209.1 | 3 | 2201 | |||
NM_019591.3 | NP_062537.2 | 4 | 2297 | |||
XM_005266182.3 | XP_005266239.1 | 5 | 2384 | |||
XM_005266183.3 | XP_005266240.1 | 6 | 2587 | |||
XM_011534829.1 | XP_011533131.1 | 5 | 2500 | |||
XM_017019920.1 | XP_016875409.1 | 6 | 3020 | |||
XM_017019921.1 | XP_016875410.1 | 6 | 2483 | |||
XM_017019922.1 | XP_016875411.1 | 5 | 3369 | |||
XM_017019923.1 | XP_016875412.1 | 5 | 3300 | |||
GenBank mRNA | AK000762.1 | - | 1 | 1500 | ||
AL833057.1 | - | 1 | 1074 | |||
BC008211.1 | - | 1 | 1037 | |||
BC046206.2 | - | 4 | 2198 | 6176427 | ||
EU668356.1 | - | 4 | 2175 |