Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033062.1 | NP_001028234.1 | 1 | 1673 | ||
GenBank mRNA | BC100630.1 | - | 1 | 1673 | 7938080 |